Learn More
Hamilton™ Bulk Pipette Tips
Delivering outstanding accuracy and precision, guaranteed.
Brand:  Hamilton™ 68008-20
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15385064
  • 81.25€ / Each
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
Description
Description
  • Standard tip length
  • Provided in bulk, resealable bags
  • Made from chemically resistant, virgin polypropylene for excellent clarity and smooth surfaces
Specifications
Specifications
200 μL
Virgin Polypropylene