Learn More
Hirschmann™ Pipetting Pliers
Product Code. 30090423
Click to view available options
:
Pliers
Product Code. 30090423
Brand: Hirschmann™ 9660101
Only null left
Add to basket
Add to basket
For the secure holding of disposable end-to-end micropipettes.
  • Especially suitable for the so-called Delbruck method, because contamination caused by direct finger contact with the outside of the capillary is avoided
  • Spreading to the reagent with falsification of analysis is reliably prevented
  • Provided in packs of 10
Specifications
Type Pliers/Tongs
For Use With (Application) Secure holding of micropipettes
Quantity 10/Pk.