Learn More
Kartell™ Pipettor tip, sterile filter, universal, in racks 1-200μL
Manufactured from polypropylene. Kartell™ Sterile Filter Tips are sterilized with gamma radiation and designed for use in molecular biology, cell cultures, bacteriology, virology, immunology, and radioimmunology. The hydrophobic PE filters act as a barrier against aerosol contamination.
Brand:  Kartell™ 0962900
Product Code. 10010822
  • 231.00€ / Pack of 960
Estimated Shipment: 27-06-2025
Explore more special offers
Add to basket
Description
Description
  • DNA-free, RNA-free, Cadium-free and are apyrogenic
  • Sterilized by gamma radiation
  • Compatibility: Kartell™, PL200, Gilson™, Eppendorf™, Socorex™, Biohit™, Nichiryo™, Brand™,Hamilton™