Learn More
Buerkle™ General Purpose Funnels PP/PE
Shop All Bürkle Products
Click to view available options
Diameter (Metric)Top:
40 mm
60 mm
80 mm
100 mm
120 mm
140 mm
160 mm
Diameter (Metric) Stem:
5 mm
8 mm
10 mm
11 mm
13 mm
16 mm
18 mm
Material:
Polyethylene
Polypropylene
Unit Size:
Each
Description
- Suitable for foodstuff
- Available in polyethylene or polypropylene
- Wide range of sizes
Specifications
Specifications
| Diameter (Metric)Top | 140 mm |
| Diameter (Metric) Stem | 16 mm |
| Material | Polyethylene |
| For Use With (Application) | For Foodstuffs |
| Includes | Suspension eyelet |
| Product Type | Funnel |