Learn More
Sampling Systems™ Plastic Powder Funnels
Shop All Sampling Systems Products
Click to view available options
Diameter (Metric)Top:
100mm
150mm
200mm
250mm
350mm
Unit Size:
Each
Description
- Manufactured from either PP or HDPE
- Crevice-free
- Available in a range of diameter sizes
Specifications
Specifications
| Diameter (Metric)Top | 250mm |
| Diameter (Metric) Stem | 50mm |
| Material | Polypropylene |
| Color | White |
| For Use With (Application) | Loading powder into containers |