Learn More
Saint-Gobain Plastic Sanitary Couplings
Click to view available options
Unit Size:
Each
Maximum Pressure:
200 psi
Description
- Reduces dead space and improves flow.
- Tapered funnel design provides a steady fluid flow.
- Smooth inner surface prevents particle entrapment.
- Seamless construction prevents contamination by eliminating leaks and areas of potential bacterial growth.
- Material meets USP Class VI requirements and is FDA compliant for food contact.
Specifications
Specifications
| Certifications/Compliance | Meets USP Class VI requirements and is FDA compliant for food contact |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| For Use With | 1/2 in. ID tubing, 1/2 in. mini clamp |
| For Use With (Application) | FDA compliant for food contact |
| Tubing Size | Fits 1/2 in. ID tubing |
| Sterilization Method | Gamma |
| Width (English) | 31/32 in. |
| Height (English) | 1 13/16 in. |