Learn More
Fisherbrand™ Nylon Filter Cloth, 300 mm x 300 mm
Product Code. 11774529 Shop All Fisherbrand Products
Click to view available options
Pore Size:
5 μm
11 μm
20 μm
30 μm
41μm
53 μm
60 μm
70 μm
100 μm
200 μm
250 μm
315 μm
400 μm
450 μm
500 μm
630 μm
700 μm
800 μm
900 μm
1000 μm
Unit Size:
Pack of 10
Product Code. 11774529
Only null left
Add to basket
Add to basket
For sieving or precision filtration of biological and chemical solutions. Fisherbrand™ Nylon Filter Cloths, 300 mm x 300 mm are available in a comprehensive range of pore sizes.
  • Operating temperature of -40° to +115°C
  • Excellent compatibility with alcohols
  • Resistant to aggressive solvents
  • Applications include solvent filtration and particle analysis
Specifications
Pore Size 900 μm
Diameter (Metric) Outer 300 mm
Frame Material Plastic
Mesh Material Nylon