Learn More
RUBIS Full Plastic Tweezers
Full Plastic Tweezers - PBT Plastic
Brand: RUBIS K2A
Additional Details : Weight : 0.00630kg
Description
- PBT Plastic
- Avoids scratches on delicate components
- Easy to use
- High Precision
- 100% Made in Switzerland
Specifications
Round, Flat | |
PBT | |
Tweezers | |
Anti-magnetic and Insulating |
4.72 in. | |
Tweezers | |
Each |