Learn More
Wuertt Celluloid™ Plasticized Drying Rack
Product Code. 11996993
Click to view available options
Height (Metric):
30 cm
61 cm
Unit Size:
Each
Product Code. 11996993
Brand: Wuertt Celluloid™ 52810111
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Benchtop rack and tray for drying laboratory glassware. Wuertt Celluloid™ Plasticized Drying Rack is constructed of plasticized wire.
  • Includes catch tray
  • Plasticized wire construction for durability
  • Angled stems
  • Looped stems for larger glassware
Specifications
Includes Drain tray
Product Type Benchtop Drainer
Holds Items (44)
Material Plasticized Wire
Positions 44
For Use With (Equipment) Laboratory glassware
No. per Pack 1 Pack
Quantity 1 Pack
Length (Metric) 42 cm
Height (Metric) 61 cm
Depth (Metric) 17 cm