Learn More
Kartell™ Plastilab™ Conical Stoppers
Features tapered conical-shaped bottom to ensure sealing action.
Brand: Kartell™ 0124100
Additional Details : Weight : 0.90000kg
Description
- Made from Polyethylene for tight and secure seal
- Raised embossed easy-to-read sizes
- Color-coded
Specifications
Conical bottom | |
Stopper | |
Polyethylene | |
DIN 12242 |
Blue | |
8.1 mm | |
NS 10/19 | |
Pentagonal top |