Learn More
Kinematica™ Plate Stand ST-P15/320 for Polytron™ 1300D Homogenizer
Product Code. 13467639 Shop All Kinematica Products
Change view
Click to view available options
For Use With (Equipment):
PT 1300 D Disperser
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
13467639 PT 1300 D Disperser Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13467639 Supplier Kinematica™ Supplier No. 11040013
Only null left
Add to basket
Add to basket
Stainless steel stand with base, height-adjustable bracket for supporting the drive and base mounting.
With height-adjustable holder for drive support stainless steel for use with Polytron™ PT 1300 D drive.
Specifications
Product Type Plate Stand
Description Stainless steel stand with base, height-adjustable bracket for supporting the drive and base mounting
For Use With (Equipment) PT 1300 D Disperser
Includes The safety ring prevents the uncontrolled lowering of the drive unit. Can also be used to fixing the drive unit height.
Model ST-P 15/320