Learn More
Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces
Product Code. 10029152
Click to view available options
Color:
Black
Clear
White
Volume (Metric) Working:
50 μL
100 μL
Product Type:
Microplate
Solid Module F8 (x12) in Frame
Solid plate 384-Well
Solid plate 96-Well
Surface Treatment:
Amine
Covalent Binding
Unit Size:
Pack of 30
Product Code. 10029152
Brand: Thermo Scientific™ 436012
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Reduce the amount of reagents used in the lab by using these plates and modules with covalent binding surfaces.
Reduce the amount of reagents used in the lab with Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces.
Covalent Surfaces (CovaLink™ and Immobilizer Amino)
  • Minimal leaching
  • Withstand vigorous washing
  • Coating with lower amounts of reagent may be possible
  • Control of orientation
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Detection Method Fluorescence
Array 16 x 24
Packaging 30 Cases, 5 Pk.
Binding Type Covalent
Color Black
Product Type Solid plate 384-Well
Material Polystyrene
Volume (Metric) Well 120 μL
No. of Wells 384
Volume (Metric) Working 50 μL
Sterility Non-sterile
Surface Treatment Amine
Total Volume (Metric) Well 120 μL
Well Shape Flat Bottom (F-well)
No. per Case 30
No. per Pack 5
Plate Blocking Unspecified
Quantity 30/Cs.
Target Molecule Peptides
Show More Show Less
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.