Learn More
Grant Instruments™ Platform for Grant Orbital Shakers and Shaker Incubators
Click to view available options
Holds:
16 x 250 to 300 mL Flasks
30 x 100 to 150 mL Flasks
6 x 1000 mL Flasks
9 x 500 mL Flasks
Unit Size:
Each
Description
- Includes clamps to keep vessels secure
- For use with
- ES-20 Shaker Incubator
- ES-80 Shaker Incubator
- PSU 10i Orbital Shaker
- PSU 20i Orbital Shaker
Specifications
Specifications
| Holds | 6 x 1000 mL Flasks |
| For Use With (Equipment) | PSU-20i Orbital Platform Shaker |
| No. of Positions | 6 |