Learn More
Platinum Cobalt Colour 50 Color Standard, Lovibond™
500mL, Undesignated, Odorless, About 3, Solution, Miscible with water in all proportions, Hexachloroplatinate of Potassium, Cobalt chloride, Hydrochloric acid
Brand: Lovibond Tintometer 134180
Additional Details : CAS : 7791-13-1 Weight : 0.50000kg Transport : UN number : 1789 Chem class : 8 Pack group : III
Description
500mL, Undesignated, Odorless, About 3, Solution, Miscible with water in all proportions, Hexachloroplatinate of Potassium, Cobalt chloride, Hydrochloric acid
Specifications
500 mL | |
Hexachloroplatinate of Potassium, Cobalt chloride, Hydrochloric acid | |
Miscible with water |
Odorless | |
∼3 | |
Solution |
Safety and Handling
- COLOUR STANDARD HAZEN 50
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid, cobalt dichloride hexahydrate