missing translation for 'onlineSavingsMsg'
Learn More

Platinum(II) acetylacetonate, 98%, Thermo Scientific Chemicals

Catalog Number p-4510688
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
15170-57-7
C10H14O4Pt
393.30
MFCD00000028
KLFRPGNCEJNEKU-FDGPNNRMSA-L
platinum ii acetylacetonate, pt acac 2, platinum bis acetylacetonate, acetylacetone platinum ii salt, platinum 2,4-pentanedionate, 2,4-pentanedione platinum ii derivative, bis acetylacetonato platinum, bis acetylacetonato platinum ii, 2,4-pentanedione, platinum ii
10960186
(Z)-4-hydroxypent-3-en-2-one;platinum
[Pt++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
15170-57-7
C10H14O4Pt
393.30
MFCD00000028
KLFRPGNCEJNEKU-FDGPNNRMSA-L
platinum ii acetylacetonate, pt acac 2, platinum bis acetylacetonate, acetylacetone platinum ii salt, platinum 2,4-pentanedionate, 2,4-pentanedione platinum ii derivative, bis acetylacetonato platinum, bis acetylacetonato platinum ii, 2,4-pentanedione, platinum ii
10960186
(Z)-4-hydroxypent-3-en-2-one;platinum
[Pt++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 250.0°C to 252.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Pt)
Packaging Glass bottle
Linear Formula [CH3COCH=C(CH3)O]2Pt
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in ethanol, diethyl ether;, soluble in acetone, toluene, diluted acids
Sublimation Point 170 °C
Formula Weight 393.31
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Platinum(II) acetylacetonate
Show More Show Less
compliance-icons
Product Identifier
  • Platinum(II) acetylacetonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'