missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Plug-In Tube Extension (Od/Id 60.3/57 mm), Straight, For Exhaust Air Ducting For U260

Use these tube extensions with Memmert ovens. Memmert&trade Plug-in Tube Extensions for Ovens are designed for exhaust air ducting and connection by hose if necessary.

Brand:  Memmert™ U260B29718

Additional Details : Weight : 0.01100kg

Product Code. 16356217

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

  • Plug-in tube extension (outer diam. 60,3 mm, inner 57 mm)
  • Angled
  • Only models U, I, S
Specifications

Specifications

Models U, UF TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 30, 55, 75, 110, 160, 260, 400, 450, 750, 1060
Plug-in Tube Extension
57 x 60.3 mm (I.D. x O.D.)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions