Learn More
Palintest™ Pocket Kit
Click to view available options
Range:
0 to 500mg/L
0 to 50mg/L
0-0.5mg/L
0-1mg/L
0-20mg/L
0-250mg/L
0-500mg/L
6.8-8.4pH
Type:
Test kit
Unit Size:
Pack of 50
Description
- Tablet reagents and accessories included in kit.
- Colorimetric test.
Specifications
Specifications
| Analytes | Organophosphate |
| Includes | All reagents and accessories |
| No. of Tests | 50 |
| Range | 0-20mg/L |
| Type | Test kit |