Learn More
Palintest™ Pocket Kit
Product Code. 11938916
Click to view available options
Range:
0 to 500mg/L
0 to 50mg/L
0-0.5mg/L
0-1mg/L
0-20mg/L
0-250mg/L
0-500mg/L
6.8-8.4pH
Type:
Test kit
Unit Size:
Pack of 50
Product Code. 11938916
Brand: Palintest™ PK131
Only null left
Add to basket
Add to basket
Conveniently packaged kit containing tablet reagents and all necessary accessories for testing based on visual color. Palintest™; Pocket Kit is inexpensive and easy to use.
  • Tablet reagents and accessories included in kit.
  • Colorimetric test.
Specifications
Analytes pH (Phenol red)
Includes All reagents and accessories
No. of Tests 50
Range 6.8-8.4pH
Type Test kit