Learn More
Palintest™ Pocket Kit
Product Code. 11938976
Click to view available options
Range:
0 to 500mg/L
0 to 50mg/L
0-0.5mg/L
0-1mg/L
0-20mg/L
0-250mg/L
0-500mg/L
6.8-8.4pH
Type:
Test kit
Unit Size:
Pack of 50
Product Code. 11938976
Brand: Palintest™ PK072
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Conveniently packaged kit containing tablet reagents and all necessary accessories for testing based on visual color. Palintest™; Pocket Kit is inexpensive and easy to use.
  • Tablet reagents and accessories included in kit.
  • Colorimetric test.
Specifications
Analytes Total Alkalinity
Includes All reagents and accessories
No. of Tests 50
Range 0-500mg/L
Type Test kit