missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ Pocket Kit

Product Code. p-8498033
Click to view available options
Range:
0 to 500mg/L
0 to 50mg/L
0-0.5mg/L
0-1mg/L
0-20mg/L
0-250mg/L
0-500mg/L
6.8-8.4pH
Type:
Test kit
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 11948916

Brand: Palintest™ PK080

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Conveniently packaged kit containing tablet reagents and all necessary accessories for testing based on visual color. Palintest™; Pocket Kit is inexpensive and easy to use.

  • Tablet reagents and accessories included in kit.
  • Colorimetric test.

Specifications

Analytes Chlorine
For Use With (Equipment) All reagents and accessories
No. of Tests 50
Range 0 to 50mg/L
Sufficient For 50 Tests
Type Test kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.