Learn More
Cole-Parmer™ PolarSafe™ Stainless Steel Liquid Nitrogen Refill Tank
Product Code. 16649561 Shop All Cole-Parmer Products
Change view
Click to view available options
Capacity (Metric):
30 L
50 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
16649561 50 L Each
16639561 30 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16649561 Supplier Cole-Parmer™ Supplier No. 4426069
Only null left
Add to basket
Add to basket
Cole-Parmer™ PolarSafe™ Stainless Steel Liquid Nitrogen Refill Tank, with self-pressurized liquid nitrogen, makes LN2 discharge safe and simple
  • Durable welded stainless steel construction ensures long-lasting performance and reduced evaporation loss
  • Unique design utilizes pressure from LN2 to discharge LN2 to other containers
  • Use as a self-pressurized LN2 backup system for cryogenic freezers
  • 5 year vacuum warranty
Specifications
Capacity (Metric) 50 L
Depth (English) 25 in.
Depth (Metric) 63.5 cm
Diameter (English) Inner 19.9 in.
Diameter (English) Outer 25 in.
Diameter (Metric) Inner 50.546 cm
Diameter (Metric) Outer 63.5 cm
Material Stainless Steel