Learn More
Palintest™ Polyacrylate Square Tube
Click to view available options
Capacity (Metric):
10 mL
Unit Size:
Pack of 5
Description
For use with Palintest™ comparator
The Palintest™ comparator with interchangeable color discs is simple to use. Just add the reagent tablet to the test sample, place in the comparator and match against the appropriate disc.
Specifications
Specifications
| Capacity (Metric) | 10 mL |
| Material | Polyacrylate |
| Packaging | 5Pack |
| Quantity | 5 Pack |
| Diameter (Metric) | 13.5 mm |
| Product Type | Test Tube |