Learn More
Kartell™ PC/PP Pipette Tips
Make pipetting safer and simple for even the most sensitive applications. Kartell™ PC/PP Pipette Tips are compatable with a wide variety of pipette models.
Brand:  Kartell™ 0962400
Additional Details : Poids : 0.80000kg
 View more versions of this product
Product Code. 10596961
  • 85.44€ / Pack of 960
En stock, prêt à être expédié : 11
Add to basket
Description
Description
  • Made of Polycarbonate/Polypropylene
  • Tip for micropipette
  • Supplied in rack
  • Compatability varies
Specifications
Specifications
Pipette tips
Natural
CE/IVD
Pipette Tip
0.5 to 10 μL
Polycarbonate/PP (Polypropylene)
Kartell™, Eppendorf™, Socorex™, Hamilton™
Rack