Learn More
Kartell™ Polycarbonate/Polypropylene Pipette Tip
Click to view available options
Volume (Metric):
100 to 1000 μL
2 to 200 μL
Unit Size:
Pack of 960
Description
- Made of Polycarbonate/Polypropylene
- Tip for micropipette
- Supplied in rack
- Compatability varies
Specifications
Specifications
| Description | Pipette tips |
| Volume (Metric) | 2 to 200 μL |
| Color | Yellow |
| Certifications/Compliance | CE/IVD |
| Tip Style | Universal |
| Packaging | Rack |
| Product Type | Pipette Tip |
| For Use With (Equipment) | Kartell, Eppendorf, Socorex, Finnpipette, Brand, Hamilton |
| Material | Polycarbonate/Polypropylene |