Learn More
Sanplatec™ Polycarbonate Vacuum Desiccator
Product Code. p-8010950
Click to view available options
Diameter (Metric) Inner:
200 mm
230 mm
Unit Size:
Each
Product Code. 11837162
Brand: Sanplatec™ 0062
Only null left
Add to basket
Add to basket
Transparent polycarbonate with excellent visibility and airtightness as well as impact resistance and heat resistance.
  • Clear view. Easy to see the condition of stored items and the timing of changing silica gel.
  • Dry seal type with O-ring. Worry-free of grease coating.
  • The highest intensity in transparent resin. Hardly broken when dropping.
  • The ceramic plate has heat resistance, corrosion resistance, and non-adhesiveness.
  • PC material feature: good pulling strength, shock strength, and heat-resistance
  • Spare plate for each type (PC-210/250 K,KG) is also available.
  • Nozzle diameter is for 9mm.
Specifications
Material Polycarbonate
Product Type Desiccator
Height (Metric) 262 mm
Height (Metric) Interior 174 mm
Diameter (Metric) Inner 200 mm
Diameter (Metric) Outer 220 mm