Learn More
Bryson Packaging™ Polyethylene Bags
Transparent and excellent for shipping, storage and as sample bags.
9.76€ - 27.14€
Product Code | Brand | Width (Metric) | Height (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Height (Metric) | Quantity | Price | Quantity & Availability | |||
12330499
|
Bryson Packaging™
BAJ-300-090U |
250 mm | 200 mm | 100 Pack |
15.70€
Pack of 100 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||
12340499
|
Bryson Packaging™
BAJ-300-110R |
300 mm | 200 mm | 50 Pack |
18.48€
Pack of 50 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||
12310499
|
Bryson Packaging™
BAJ-300-050J |
100 mm | 150 mm | 100 Pack |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12350499
|
Bryson Packaging™
BAJ-300-130L |
375 mm | 250 mm | 50 Pack |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
Description
- Available in a wide range of sizes/capacities
- Suitable for food use
Specifications
Bag | |
Polyethylene |