Learn More
Masterflex™ Polyethylene Barbed Elbow Connector
Create ninety-degree bends in flexible tubing without the kinks
24.85€ - 31.40€
Specifications
Product Type Barbed Elbow Connector
Description Barbed Connection
Autoclavable Not Autoclavable
Material HDPE
Sterilization Method Ethylene Oxide
View More Specs
Products 2
Product Code Brand Tubing Size Price Quantity & Availability  
Product Code Brand Tubing Size Price Quantity & Availability  
13556720
View Documents Promotion Details
Masterflex™
30622-67
3/4 in.
31.40€
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
11919668
View Documents Promotion Details
Masterflex™
30622-72
3/16 in.
24.85€
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
Description
Description
  • Ideal for low-pressure applications
  • Chemical resistant
  • Available in a variety of sizes
  • Economical barbed fittings easily connect fluid paths without the use of tools
  • Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifications
Specifications
Barbed Elbow Connector
Not Autoclavable
Ethylene Oxide
Barbed Connection
HDPE
Translucent