Learn More
Masterflex™ Polyethylene Barbed Y-Connectors
Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing
Brand:  Masterflex™ 30726-21
Additional Details : Poids : 0.04000kg
 View more versions of this product
Product Code. 15347349
  • 7.35€ / Pack of 10
Expédition estimée
29-10-2020
Add to basket
Description
Description
  • Barbed Y-Connectors
  • Polyethylene
  • Available in pack of ten
Specifications
Specifications
Translucent
Not Autoclavable
Barbed Y-Connector
1/16 in.
1/16 in.
Barb Connections
HDPE
1/16 in.
1/16 in.
Ethylene Oxide