Learn More
Dueperthal™ Polyethylene Brush Container
Click to view available options
Capacity (Metric):
0.45 L
0.9 L
1.5 L
Unit Size:
Each
Specifications
Specifications
| Capacity (Metric) | 0.45 L |
| Color | White |
| Height (Metric) | 150 mm |
| Includes | Cap, Brush |
| Material | Polyethylene |
| Product Type | Brush Container |