Learn More
Buerkle™ Polyethylene Bung
Manufactured with polyethylene for strength, durability and flexibility. Buerkle™ Polyethylene Bungs are available in a wide range of sizes.
Brand: Buerkle™ 9760-2932
This item is not returnable.
View return policy
Description
- Hollow stopper allows for use of tubes or funnels
- Sealing rings provide a tight seal
- Suitable for foodstuff
- Octagonal shape facilitates easy opening
- Polyethylene provides good chemical resistance