Learn More
Kartell™ Polyethylene Disc Membrane
Click to view available options
Diameter (Metric):
45 mm
55 mm
70 mm
80 mm
90 mm
110 mm
160 mm
240 mm
Unit Size:
Pack of 10
Description
- Useful interface between filter paper and perforated Buchner plate
- Avoids clogging of filter holes
Specifications
Specifications
| Filter Type | Screen Filter |
| Format | Disc |
| Material | Polyethylene |
| Pore Size | 300 μm |
| For Use With (Application) | Interface between filter paper and perforated Buchner plate |
| No. per Pack | 10/Pack |
| Shape | Circle |
| Diameter (Metric) | 110 mm |