missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Polyethylene Flat Seal

Product Code. 30276786 Shop All Bürkle Products
Click to view available options
Includes:
2 in. thread
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 30276786

Brand: Buerkle™ 85380200

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Flat seals made of Polyethylene, transparent fitting outer or inner thread.

  • PE
TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
For Use With (Application) For Use with Foodstuffs
Includes 2 in. thread
Quantity 10 Pk.
Material Polyethylene
Type Seal, Flat
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.