missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol 200 Grade, Extra Pure, SLR, Fisher Chemical™

Brand:  Fisher Chemical P/3675/25

Additional Details : CAS : 25322-68-3 Weight : 25.00000kg


Packaging Plastic container
Quantity 25L

 View more versions of this product

Product Code. 12678645

  • 394.00€ / 25 L


In stock, ready to ship: 5
Add to basket

Description

Description

Specifications

Chemical Identifiers

25322-68-3
81839

Specifications

25322-68-3
200g/mol
Plastic container
60 to 70 mPaS at 20°C
5.0 to 7.0
81839
Liquid
<0.01mmHg at 20°C
-65°C
25L
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Polyethylene glycol - 200 grade

Signal Word
  • Warning

Hazard Category
  • Specific target organ toxicity Category 3

Hazard Statement
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS Documents