Learn More
Bel-Art™ Scienceware™ Polyethylene Magnetic Stirring Bar Retriver
Product Code. 11785064 Shop All SP Scienceware Products
Click to view available options
Material:
HDPE
Type:
Magnetic Stir Bar Retriever
Unit Size:
Each
Product Code. 11785064
Brand: Bel-Art™ Scienceware™ F377750000
Only null left
Add to basket
Add to basket
Eliminates contamination of contents Does not need to be cleaned or autoclaved after use Magnet is located in the 25 x 70mm rectangular section; overall length: 20cm (7.7 in.) Long ribbed handle provides a secure grip Made of high density polyethylene
  • Eliminates contamination of contents
  • Does not need to be cleaned or autoclaved after use 
  • Magnet is located in the 25 x 70mm rectangular section; overall length: 20cm (7.7”)
  • Long ribbed handle provides a secure grip
  • Made of high density polyethylene
Specifications
Material HDPE
Type Magnetic Stir Bar Retriever