Learn More
Masterflex™ Polyethylene Male Pipe Adapter
Click to view available options
Thread Size:
1/2 x 3/4 in.
1/2 x 5/8 in.
1/4 x 1/2 in.
1/4 x 5/16 in.
1/8 x 1/8 in.
3/4 x 3/4 in.
Unit Size:
Pack of 10
Description
- Polyethylene Male Pipe Adapter
- Adapts from a NPT male thread to a hose barb
- Barbed fittings are designed for quick and easy assembly without tools
- Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
- Available in packs of ten
Specifications
Specifications
| Description | Barb x NPT(M) Connection |
| Autoclavable | Not Autoclavable |
| Material | HDPE |
| Packaging Quantity | 10 Fittings |
| Tubing Size | 5/8 in. |
| Outlet Port | 1/2 in. |
| Port Size | 5/8 in. |
| Sterilization Method | Ethylene Oxide |
| Thread Size | 1/2 x 5/8 in. |
| Shape | Straight |
| Show More |