missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Plastic Snap Caps, N22

Product Code. 10717552 Shop All Macherey-Nagel Products
Click to view available options
Color:
Transparent
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10717552

Brand: Macherey-Nagel™ 70275

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Used for the storage of powdery samples. Macherey-Nagel™ Plastic Snap Caps, N22 are transparent polyethylene caps for use with Macherey-Nagel™ Glass Snap Cap Vials, N22. They feature a small tab so they can be easily popped off. Packs of 100 in PE bags.

All products are manufactured under strict hygienic conditions. In-process controls and a final QC inspection are also part of the measurements that have been implemented to grant a reliable product quality.

Specifications

Diameter (Metric) Cap 22 mm
Color Transparent
Material Polyethylene
For Use With (Application) Storage of powdery samples
Quantity 100 Pk.
Type Snap Cap
Closure Size N22
For Use With (Equipment) Glass Snap Cap Vials, N22 (702019 and 70273)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.