Learn More
Masterflex™ Polyethylene Pipe Adapter Elbow Connector Barbed Fitting
Product Code. 11759859
Click to view available options
For Use With:
1/4 NPT(M) x 1/4 in. tubing I.D.
1/4 NPT(M) x 3/8 in. tubing I.D.
Unit Size:
Pack of 10
Product Code. 11759859
Brand: Masterflex™ 3062298
Only null left
Specifications
Material Polyethylene
For Use With 1/4 NPT(M) x 3/8 in. tubing I.D.
Packaging Quantity 10 Pack
Thread Size 1/4 NPT(M)
Includes Pipe adapter Elbow connector; ETO sterilization
Product Type Barbed Fitting