Learn More
Buerkle™ Polyethylene Round All Purpose Boxes
Product Code. 10382292 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Product Code. 10382292
Brand: Buerkle™ 62102000
Box for storing, packing, taking samples, sampling and transport of pastes, granules, powders, soil samples and frozen goods (-40 °C up to briefly +95 °C). Ideal use of space, can be stacked. Complete with press-in lid
  • PE, transparent
  • No crevices
  • |PE, transparent
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 2000 mL
Color Transparent
Material Polyethylene
Shape Round
For Use With (Application) For Storing, Packing, Taking Samples, Sampling and Transport of Pastes, Granules, Powders, Soil Samples and Frozen Goods (-40 up to Briefly 95°C)
Product Type All purpose box
Height (Metric) 143 mm
Diameter (Metric) Outer 169 mm (top), 148 mm (bottom)