Learn More
Gravis Polyethylene Snap-Caps
Click to view available options
Thread Size (Metric):
22 mm
30 mm
40 mm
Unit Size:
Pack of 100
Pack of 86
Description
- Durable
- Provides excellent chemical resistance
Specifications
Specifications
| Material | Polyethylene |
| No. per Pack | 86Pack |
| Quantity | 86 Pack |
| Thread Size (Metric) | 40 mm |
| Product Type | Snap Cap |
| Closure Type | Snap |