missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Polyethylene Wide Mouth Two Color Safety Label Wash Bottle
Click to view available options
Chemical Label:
Acetone
Ethyl Acetate
Methanol
Water
Description
Wide-Mouth wash bottles for easy refilling.
- Valve eliminates drips — allows dispensing of wash bottle in an upright or inverted position
- Wide mouth 53mm (2 in.) bottle for easy refilling
- Polypropylene closure; color-coded to aid in easy identification
- Translucent, LDPE solvent-resistant
- Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Specifications
Closure Material | Polypropylene |
Closure Size | 53 mm |
Closure Type | Green Polypropylene Cap |
Through the Closure Dispensing | Yes |
Chemical Label | Ethyl Acetate |
Product Type | Wash Bottle |
Capacity (Metric) | 500 mL |
Capacity (English) | 16.9 oz. |
Color | Translucent |
Diameter (Metric) Outer Neck | 53mm |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'