missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygoprep™ 60 to 80

Brand:  Macherey-Nagel™ 711270.1000

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10012731

  • 190.00€ / 1000g
Estimated Shipment: 07-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

63 to 100 μm
POLYGOPREP 60 to 80
2 to 8
POLYGOPREP
Polygoprep
Liquid Chromatography
60 Å
Liquid Chromatography
1 kg
350 m2/g
Silica
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions