Learn More
Macherey-Nagel™ Alox-25 UV254 TLC Glass Plates
Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
5 x 20 cm
20 x 20 cm
Unit Size:
Pack of 100
Pack of 25
Specifications
Specifications
| Pore Size | 60 Å |
| Quantity | 100 Pk. |
| Type | Glass Plates |
| Fluorescent Indicator | UV254 |
| Thickness (Metric) | 0.25 mm |
| Dimensions (L x W) | 5 x 20 cm |