Learn More
Macherey-Nagel™ Polygram CEL 400 UV254 Polyester Sheets
Product Code. 10556711 Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
20 x 20 cm
Unit Size:
Pack of 25
Product Code. 10556711
Brand: Macherey-Nagel™ 801123
Only null left
Add to basket
Add to basket
TLC polyester sheets with fibrous cellulose impregnated with polyethyleneimine (PEI).
Specifications
Quantity 25 Pk.
Type Polyester Sheet
Fluorescent Indicator UV254
Thickness (Metric) 0.1 mm
Dimensions (L x W) 20 x 20 cm