Learn More
Macherey-Nagel™ Polygram IONEX-25 SA-Na Polyester Sheets
Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
20 x 20 cm
Unit Size:
Pack of 25
Specifications
Specifications
| Quantity | 25 Pk. |
| Type | Polyester Sheet |
| Thickness (Metric) | 0.2 mm |
| Dimensions (L x W) | 20 x 20 cm |