Learn More
Kimberly-Clark™ KLEENGUARD™ A10 Light Duty Coveralls with Hood
Offering CE Simple (minimal risk) protection for the user engaged in light duty tasks and basic contamination control. Kimberly-Clark™ KLEENGUARD™ A10 Light Duty Coveralls with Hood are manufactured from spunbond polypropylene material. 
Brand:  Kimberly-Clark™ 95670
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12681476
  • 258.00€ / Pack of 50
Estimated Shipment
24-03-2021
Add to basket
Description
Description
  • Silicone-free and individually packed
  • Reduces the risk of heat stress
  • Easy fastening snap front
  • Triple-stitched seams to help protect against tearing
  • Offers complete freedom of movement with comfortable body design and elasticated cuffs, ankles and waist
Specifications
Specifications
2X-Large
7.98 kg