Learn More
KleenGuard™ A71 Chemical Spray Protection Coveralls
Product Code. p-8005923
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 10
Product Code. 12631396
Brand: KleenGuard™ 96770
Only null left
Add to basket
Add to basket
Coveralls create a spray-tight and liquid-tight barrier against harmful substances when working with hazardous chemicals.
  • Approved as Complex Design (Category 3) equipment - offering protection to the levels specified for Type 3, 4 and 5 by CEN
  • Highly visible yellow color - for improved worker safety
  • Ideal for working with hazardous chemicals - protecting against liquid, aerosol and dry chemical splashes, sprays and dusts in low-pressure industrial cleaning and maintenance areas
  • Single-use protective clothing to improve safety and comfort in high-risk areas - create a spray and liquid tight barrier against harmful chemical splashes, liquids or dust particles
  • Suitable for use as protection against Infective Agents and Biohazards
  • Suitable for use as protection against solid particles and particulate radioactive contamination
  • Treated to be antistatic
  • EN 1073-2 approved for radioactive dust protection
  • Anti-static EN 1149-1 for critical areas
Specifications
Product Type General Purpose Coveralls
Size Large
Material Spunbond Polypropylene
Color Yellow
Disposable Reusable
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Protection Class Type 3, 4 and 5
Resistance Perforation, Flame, Abrasion, Tears, Particle, Liquid
Latex Free Yes
No. of Pockets None
For Use With (Application) Hazardous Chemicals
Style Full Body