missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Polypropylene Ball Valve, Food

Product Code. 17240674 Shop All Bürkle Products
Click to view available options
Width (Metric):
20 mm
25 mm
N/A
Outlet Port:
M29 x 1,75, Outer
M36 x 3, Outer
M42 x 3, Outer
N/A
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17240674

Brand: Buerkle™ 05008202

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ball valve made of PP with outer / inner thread, fibre-glass reinforced, ball made of PP, handle with O-ring made of food-safe NBR and seal made of HDPE.

  • Max. operating pressure 1 bar (at +20 °C)
  • Max. operating temperature 100 °C
  • Suitable for food
TRUSTED_SUSTAINABILITY

Specifications

Type Ball Valve
Material Polypropylene
Width (Metric) N/A
No. per Pack 1 Ea.
For Use With (Equipment) Thread, Inflowing Side: 0.5 in. Inner; Thread Outflowing Side: M29 x 1,75, Outer
Outlet Port M29 x 1,75, Outer
Color White and red
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.