missing translation for 'onlineSavingsMsg'
Learn More

Semadeni™ Color-Coded Plastic Beakers

Product Code. 15829465
Click to view available options
Color:
Blue
Green
Natural
Yellow
Unit Size:
Each
Pack of 100
This item is not returnable. View return policy

Product Code. 15829465

Brand: Semadeni™ 8613

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Excellent for the storage, mixing or pouring of liquids in the laboratory. Semadeni™ Color-Coded Plastic Beakers are manufactured from durable polypropylene.

  • Translucent colors
  • 30mL capacity
  • Graduated
  • Optional LDPE lids available for purchase

Specifications

Color Green
Capacity (Metric) 30 mL
Material Polypropylene
For Use With (Application) Suitable as dosing or laboratory beakers
Type Beaker
Disposable Single-use
Graduated Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.