Learn More
Sigma Laborzentrifugen™ Polypropylene Bottle
A durable vessel for use with Sigma™ centrifuge carriers.
Brand:  Sigma Laborzentrifugen™ 15931
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 13568780
  • 211.00€ / Each
Estimated Shipment
05-06-2020
Add to basket
Specifications
Specifications
1500 mL
180mm
Yes
115mm
Polypropylene
Centrifuge Bottle
13850 with 17175
Screw cap