missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wheaton™ Polypropylene Cap
Shop All Wheaton Science Products Products
Click to view available options
Neck Finish:
20-400
33-400
45-400
63-400
Unit Size:
Pack of 144
Pack of 300
Pack of 48
Pack of 72
Specifications
Specifications
| Color | White |
| For Use With (Application) | Storage and Retrieval of Volatile Solvents/Sensitive Compounds and Corrosive Chemicals |
| Material | Polypropylene |
| Liner Material | Polyethylene Foam Liner |
| Neck Finish | 33-400 |
| Product Type | Cap |
| Autoclavable | Autoclavable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction